Assay Details
Target Gene Details
Entrez Gene ID: | 72795 |
Gene Name: | tetratricopeptide repeat domain 19 |
Gene Aliases: |
2010204O13Rik, 2810460C24Rik, AI505442 |
Location: |
Chr.11:62281427-62316441 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc19 | NM_028360.2 | NP_082636.3 | ||
| NM_029704.2 | NP_083980.2 | |||
| AK013368.1 | BAB28813.2 | |||
| AK034075.1 | BAC28572.1 | |||
| BC026866.1 | AAH26866.1 | |||
| BC037111.1 | AAH37111.1 |

Genomic Map