Assay Details
Target Gene Details
Entrez Gene ID: | 55943 |
Gene Name: | syntaxin 8 |
Gene Aliases: |
0610007H08Rik, 1110002H11Rik, 4930571E13Rik, CARB |
Location: |
Chr.11:67966411-68207148 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stx8 | NM_018768.2 | NP_061238.1 | ||
| AB040054.1 | BAB20500.1 | |||
| AF036716.1 | AAC95286.1 | |||
| AF093064.1 | AAC64149.1 | |||
| AK003297.1 | BAB22698.1 | |||
| AK011376.1 | BAB27578.1 | |||
| AK016271.1 | ||||
| AK018716.1 | ||||
| BC048167.1 | AAH48167.1 | |||
| BC048479.1 | AAH48479.1 | |||
| BC061118.1 | AAH61118.1 |
More Information
Set Membership: |
|

Genomic Map