Assay Details
Target Gene Details
Entrez Gene ID: | 20913 |
Gene Name: | syntaxin binding protein 4 |
Gene Aliases: |
6030470M02Rik, Synip |
Location: |
Chr.11:90476492-90638123 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stxbp4 | NM_011505.2 | NP_035635.1 | ||
| AF152924.1 | AAD43533.1 | |||
| AK012293.1 | ||||
| AK049001.1 | ||||
| BC032881.1 | AAH32881.1 |

Genomic Map