Assay Details
Target Gene Details
Entrez Gene ID: | 21833 |
Gene Name: | thyroid hormone receptor alpha |
Gene Aliases: |
6430529J03Rik, AW259572, Erba, Nr1a1, Rvr, T3R[a], T3Ralpha, Thra1, Thra2 |
Location: |
Chr.11:98741873-98765111 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Thra | NM_178060.3 | NP_835161.1 | ||
| AK078233.1 | BAC37186.1 | |||
| AK082121.1 | ||||
| AK192370.1 | ||||
| X07750.1 | CAA30575.1 | |||
| X51983.1 | CAA36241.1 |
More Information
Set Membership: |
|

Genomic Map