Assay Details
Target Gene Details
Entrez Gene ID: | 170930 |
Gene Name: | SMT3 suppressor of mif two 3 homolog 2 (yeast) |
Gene Aliases: |
SUMO-2, Smt3A, Smt3b, Smt3h2 |
Location: |
Chr.11:115523109-115536230 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sumo2 | NM_133354.2 | NP_579932.1 | ||
| AK012619.1 | BAB28360.1 | |||
| AK085238.1 | BAC39397.1 | |||
| AK160404.1 | BAE35772.1 | |||
| AK160418.1 | ||||
| AK167308.1 | BAE39412.1 | |||
| AK219519.1 | ||||
| BC017522.1 | AAH17522.1 | |||
| BC083326.1 | AAH83326.1 | |||
| BC095930.1 | AAH95930.1 | |||
| L79948.1 | AAL40136.1 |

Genomic Map