Assay Details
Target Gene Details
Entrez Gene ID: | 21927 |
Gene Name: | tumor necrosis factor, alpha-induced protein 1 (endothelial) |
Gene Aliases: |
Edp-1, Edp1, Tnfip1 |
Location: |
Chr.11:78522850-78536260 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfaip1 | NM_001159392.1 | 3 | 537 | NP_001152864.1 |
| NM_009395.4 | 3 | 550 | NP_033421.3 | |
| AK004593.2 | 3 | 464 | BAB23395.1 | |
| AK034305.1 | 3 | 485 | BAC28668.1 | |
| AK147809.1 | 3 | 445 | BAE28152.1 | |
| AK168386.1 | 3 | 429 | BAE40313.1 | |
| AK168704.1 | 3 | 472 | BAE40546.1 | |
| AK168805.1 | 3 | 445 | ||
| AK172287.1 | BAE42927.1 | |||
| AK216341.1 | 2 | 107 | ||
| BC003906.1 | 3 | 386 | AAH03906.1 |
More Information
Set Membership: |
|

Genomic Map