Assay Details
Target Gene Details
Entrez Gene ID: | 217154 |
Gene Name: | SH3 and cysteine rich domain 2 |
Gene Aliases: |
24b2, 24b2/STAC2, AW240854 |
Location: |
Chr.11:98036624-98053462 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stac2 | NM_146028.4 | NP_666140.1 | ||
| AJ608761.1 | CAE75538.1 | |||
| AK078132.1 | BAC37142.1 | |||
| AK079357.1 | BAC37621.1 | |||
| AK144433.1 | BAE25886.1 | |||
| BC024864.1 | AAH24864.1 | |||
| BC029794.1 | AAH29794.1 | |||
| BC054100.1 | AAH54100.1 |
More Information
Set Membership: |
|

Genomic Map