Assay Details
Target Gene Details
Entrez Gene ID: | 104776 |
Gene Name: | aldehyde dehydrogenase family 6, subfamily A1 |
Gene Aliases: |
1110038I05Rik, AI314632, Mmsdh |
Location: |
Chr.12:84430717-84450950 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aldh6a1 | NM_134042.2 | NP_598803.1 | ||
| AF297860.1 | AAG44988.1 | |||
| AK033587.1 | BAC28375.1 | |||
| AK082790.1 | ||||
| AK128924.1 | ||||
| AK147146.1 | BAE27715.1 | |||
| AK155814.1 | BAE33445.1 | |||
| AK164366.1 | ||||
| AK169915.1 | BAE41455.1 | |||
| AK170305.1 | BAE41702.1 | |||
| AK171581.1 | BAE42539.1 | |||
| AK171896.1 | BAE42726.1 | |||
| BC031148.1 | AAH31148.1 | |||
| BC033440.1 | AAH33440.1 | |||
| BC115855.1 |
More Information
Set Membership: |
|

Genomic Map