Assay Details
Target Gene Details
Entrez Gene ID: | 20926 |
Gene Name: | suppressor of Ty 6 |
Gene Aliases: |
5131400N11Rik, AI132449, SPT6, Supt6h, mKIAA0162 |
Location: |
Chr.11:78206749-78245703 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Supt6 | NM_009297.2 | 18 | 2447 | NP_033323.2 |
| AK129069.1 | 6 | 770 | BAC97879.1 | |
| AK170533.1 | 18 | 2446 | ||
| BC072657.1 | 18 | 2367 | AAH72657.1 | |
| U40375.1 | 18 | 2445 | AAB18950.1 |
More Information
Set Membership: |
|

Genomic Map