Assay Details
Target Gene Details
Entrez Gene ID: | 22627 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, epsilon polypeptide |
Gene Aliases: |
AU019196 |
Location: |
Chr.11:75732887-75765841 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ywhae | NM_009536.4 | 1 | 56 | NP_033562.3 |
| AK075995.1 | BAC36106.1 | |||
| AK135831.1 | BAE22681.1 | |||
| AK135867.1 | BAE22705.1 | |||
| AK169812.1 | BAE41384.1 | |||
| BC058686.1 | AAH58686.1 | |||
| BY293945.1 | 1 | 56 | ||
| Z19599.1 | CAA79659.1 |
More Information
Set Membership: |
|

Genomic Map