Assay Details
Target Gene Details
Entrez Gene ID: | 22627 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, epsilon polypeptide |
Gene Aliases: |
AU019196 |
Location: |
Chr.11:75732887-75765841 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ywhae | NM_009536.4 | 2 | 299 | NP_033562.3 |
| AF483478.1 | 2 | 158 | AAL90752.1 | |
| AF483479.1 | 2 | 158 | AAL90753.1 | |
| AK075995.1 | 2 | 248 | BAC36106.1 | |
| AK135831.1 | 2 | 227 | BAE22681.1 | |
| AK135867.1 | 2 | 227 | BAE22705.1 | |
| AK169812.1 | 2 | 254 | BAE41384.1 | |
| AK191470.1 | 1 | 57 | ||
| AK192964.1 | 1 | 57 | ||
| AK202856.1 | 1 | 57 | ||
| AK212566.1 | 1 | 57 | ||
| AK214161.1 | 1 | 57 | ||
| AK218792.1 | 1 | 57 | ||
| AK220088.1 | 1 | 57 | ||
| AX775991.1 | 2 | 302 | CAE11695.1 | |
| AX775993.1 | 1 | 95 | CAE11696.1 | |
| BC058686.1 | 2 | 258 | AAH58686.1 | |
| BY293945.1 | 2 | 299 | ||
| D87663.1 | 2 | 186 | BAA13424.1 | |
| Z19599.1 | 2 | 241 | CAA79659.1 |
More Information
Set Membership: |
|

Genomic Map