Assay Details
Target Gene Details
Entrez Gene ID: | 22627 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, epsilon polypeptide |
Gene Aliases: |
AU019196 |
Location: |
Chr.11:75732887-75765841 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ywhae | NM_009536.4 | NP_033562.3 | ||
| AF483478.1 | AAL90752.1 | |||
| AF483479.1 | AAL90753.1 | |||
| AK075995.1 | BAC36106.1 | |||
| AK135831.1 | BAE22681.1 | |||
| AK135867.1 | BAE22705.1 | |||
| AK169812.1 | BAE41384.1 | |||
| AK179917.1 | ||||
| AK183177.1 | ||||
| AK188179.1 | ||||
| AK188405.1 | ||||
| AK190793.1 | ||||
| AK191008.1 | ||||
| AK192790.1 | ||||
| AK192964.1 | ||||
| AK195324.1 | ||||
| AK195513.1 | ||||
| AK204268.1 | ||||
| AK207068.1 | ||||
| AK218242.1 | ||||
| AX775991.1 | CAE11695.1 | |||
| AX775993.1 | CAE11696.1 | |||
| BC058686.1 | AAH58686.1 | |||
| D87663.1 | BAA13424.1 | |||
| Z19599.1 | CAA79659.1 |
More Information
Set Membership: |
|

Genomic Map