Assay Details
Target Gene Details
Entrez Gene ID: | 217732 |
Gene Name: | CLOCK interacting protein, circadian |
Gene Aliases: |
2310044G17Rik, AI842544, AI853744, Kiaa1737, mKIAA1737 |
Location: |
Chr.12:86947041-86965364 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cipc | NM_001289429.1 | NP_001276358.1 | ||
| NM_001289430.1 | NP_001276359.1 | |||
| NM_001289431.1 | 2 | 388 | NP_001276360.1 | |
| NM_001289432.1 | NP_001276361.1 | |||
| NM_173735.3 | NP_776096.2 | |||
| AK122546.1 | BAC65828.1 | |||
| AK148415.1 | BAE28540.1 | |||
| AK154476.1 | BAE32612.1 | |||
| AK160481.1 | ||||
| AK160643.1 | BAE35935.1 | |||
| AK165964.1 | 2 | 283 | ||
| AK210327.1 | ||||
| BC026384.1 | AAH26384.1 | |||
| BY097396.1 | ||||
| BY287209.1 | ||||
| CF530959.1 | ||||
| CK391711.1 | 2 | 260 | ||
| CN526809.1 |

Genomic Map