Assay Details
Target Gene Details
Entrez Gene ID: | 268567 |
Gene Name: | transmembrane protein 229B |
Gene Aliases: |
6330442E10Rik, 9830138K24, AU019070 |
Location: |
Chr.12:78961795-79007659 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem229b | NM_001170401.1 | NP_001163872.1 | ||
| NM_178745.4 | NP_848860.1 | |||
| AK033403.1 | BAC28272.1 | |||
| AK036593.1 | BAC29498.1 | |||
| AK053623.1 | BAC35450.1 | |||
| AK076323.1 | 1 | 2327 | ||
| AK151405.1 | BAE30372.1 | |||
| AK169759.1 | BAE41349.1 | |||
| AK211134.1 | ||||
| BC066067.1 | AAH66067.1 | |||
| BC079613.1 | AAH79613.1 |

Genomic Map