Assay Details
Target Gene Details
Entrez Gene ID: | 104111 |
Gene Name: | adenylate cyclase 3 |
Gene Aliases: |
AC3, mKIAA0511 |
Location: |
Chr.12:4133087-4213525 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adcy3 | NM_001159536.1 | NP_001153008.1 | ||
| NM_001159537.1 | NP_001153009.1 | |||
| NM_138305.3 | NP_612178.2 | |||
| AF458089.1 | AAL59384.1 | |||
| AK122298.1 | BAC65580.1 | |||
| AK191459.1 | ||||
| AK218847.1 | ||||
| BC057113.1 | AAH57113.1 | |||
| BC057316.1 | AAH57316.1 |

Genomic Map