Assay Details
Target Gene Details
Entrez Gene ID: | 52615 |
Gene Name: | suppressor of zeste 12 homolog (Drosophila) |
Gene Aliases: |
2610028O16Rik, AI195385, AU016842, AW536442, D11Ertd530e, mKIAA0160 |
Location: |
Chr.11:79993106-80034123 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Suz12 | NM_001163018.1 | 11 | 1520 | NP_001156490.1 |
| NM_199196.2 | 12 | 1589 | NP_954666.1 | |
| AK122213.1 | 12 | 1574 | BAC65495.1 | |
| AK135897.1 | 6 | 635 | BAE22718.1 | |
| AK141264.1 | 3 | 193 | BAE24622.1 | |
| BC051099.1 | 3 | 283 | AAH51099.1 | |
| BC064461.1 | 12 | 1589 | AAH64461.1 | |
| BC084591.1 | 7 | 860 | AAH84591.1 |
More Information
Set Membership: |
|

Genomic Map