Assay Details
Target Gene Details
Entrez Gene ID: | 52615 |
Gene Name: | suppressor of zeste 12 homolog (Drosophila) |
Gene Aliases: |
2610028O16Rik, AI195385, AU016842, AW536442, D11Ertd530e, mKIAA0160 |
Location: |
Chr.11:79993106-80034123 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Suz12 | NM_001163018.1 | NP_001156490.1 | ||
| NM_199196.2 | NP_954666.1 | |||
| AK122213.1 | BAC65495.1 | |||
| AK141264.1 | BAE24622.1 | |||
| AK207341.1 | ||||
| BC064461.1 | AAH64461.1 | |||
| W13031.1 |
More Information
Set Membership: |
|

Genomic Map