Assay Details
Target Gene Details
Entrez Gene ID: | 104111 |
Gene Name: | adenylate cyclase 3 |
Gene Aliases: |
AC3, mKIAA0511 |
Location: |
Chr.12:4133087-4213525 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adcy3 | NM_001159536.1 | 7 | 1725 | NP_001153008.1 |
| NM_001159537.1 | 7 | 1784 | NP_001153009.1 | |
| NM_138305.3 | 6 | 1402 | NP_612178.2 | |
| AF253540.1 | 3 | 273 | AAF65474.1 | |
| AF458089.1 | 6 | 1374 | AAL59384.1 | |
| AK122298.1 | 7 | 1696 | BAC65580.1 | |
| BC057113.1 | 7 | 1713 | AAH57113.1 | |
| BC057316.1 | 7 | 1614 | AAH57316.1 |

Genomic Map