Assay Details
Target Gene Details
Entrez Gene ID: | 93670 |
Gene Name: | tachykinin 4 |
Gene Aliases: |
AW489379, PPT-C |
Location: |
Chr.11:95261529-95269262 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tac4 | NM_053093.1 | NP_444323.1 | ||
| AF235035.1 | AAK15025.1 | |||
| AF515827.1 | AAP30873.1 | |||
| BC119426.1 |
More Information
Set Membership: |
|

Genomic Map