Assay Details
Target Gene Details
Entrez Gene ID: | 21414 |
Gene Name: | transcription factor 7, T cell specific |
Gene Aliases: |
AI465550, TCF-1, Tcf1 |
Location: |
Chr.11:52252283-52283378 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tcf7 | NM_009331.3 | NP_033357.1 | ||
| AK037616.1 | BAC29835.1 | |||
| BC138596.1 | ||||
| BC145343.1 | ||||
| X61385.1 | CAA43658.1 |
More Information
Set Membership: |
|

Genomic Map