Assay Details
Target Gene Details
Entrez Gene ID: | 79221 |
Gene Name: | histone deacetylase 9 |
Gene Aliases: |
AV022454, D030072B18Rik, HD7B, HD9, HDRP, Hdac7b, Mitr, mKIAA0744 |
Location: |
Chr.12:34047580-34917095 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hdac9 | NM_001271386.1 | NP_001258315.1 | ||
| NM_024124.3 | NP_077038.2 | |||
| AF235053.1 | AAK15027.1 | |||
| AF279371.1 | AAL86358.1 | |||
| AF324492.1 | AAG48332.1 | |||
| AK087563.1 | ||||
| AK133334.1 | ||||
| BC098187.1 | AAH98187.1 | |||
| EL604734.1 |
More Information
Set Membership: |
|

Genomic Map