Assay Details
Target Gene Details
Entrez Gene ID: | 71091 |
Gene Name: | cyclin-dependent kinase-like 1 (CDC2-related kinase) |
Gene Aliases: |
4933411O17Rik |
Location: |
Chr.12:69746846-69791219 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdkl1 | NM_183294.2 | NP_899117.1 | ||
| AK016781.1 | BAC25497.1 | |||
| BC115909.1 |
More Information
Set Membership: |
|

Genomic Map