Assay Details
Target Gene Details
Entrez Gene ID: | 23876 |
Gene Name: | fibulin 5 |
Gene Aliases: |
A55, DANCE, EVEC |
Location: |
Chr.12:101746565-101819130 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbln5 | NM_011812.4 | 10 | 1454 | NP_035942.1 |
| AF112151.1 | 10 | 1404 | AAD41767.1 | |
| AK085170.1 | 10 | 1284 | BAC39381.1 | |
| AK090129.1 | 10 | 1393 | BAC41109.1 | |
| AK159189.1 | 10 | 1218 | ||
| AK159471.1 | 10 | 1437 | BAE35112.1 | |
| AK159960.1 | 10 | 1424 | BAE35515.1 | |
| AK165018.1 | 10 | 1247 | BAE38002.1 | |
| AK184082.1 | 1 | 95 | ||
| AK192931.1 | 1 | 95 | ||
| AK201795.1 | 1 | 95 | ||
| AK201994.1 | 1 | 95 | ||
| AK203142.1 | 1 | 95 | ||
| AK205213.1 | 1 | 95 | ||
| AK219436.1 | ||||
| BC006636.1 | 10 | 1144 | AAH06636.1 |
More Information
Set Membership: |
|

Genomic Map