Assay Details
Target Gene Details
Entrez Gene ID: | 207965 |
Gene Name: | valosin containing protein lysine (K) methyltransferase |
Gene Aliases: |
Gm71, Mettl21d, Mettl221d, VCP-KMT |
Location: |
Chr.12:69576730-69583109 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vcpkmt | NM_001033236.2 | NP_001028408.2 | ||
| AK083148.1 | 1 | 1804 | BAC38784.1 | |
| AK144832.1 | BAE26088.1 | |||
| BC138942.1 | ||||
| BC138943.1 |

Genomic Map