Assay Details
Target Gene Details
Entrez Gene ID: | 58208 |
Gene Name: | B cell leukemia/lymphoma 11B |
Gene Aliases: |
9130430L19Rik, AI604821, B630002E05Rik, BCL-11B, Ctip2, Rit1 |
Location: |
Chr.12:107910403-108004139 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcl11b | NM_001079883.1 | 3 | 770 | NP_001073352.1 |
| NM_001286343.1 | NP_001273272.1 | |||
| NM_021399.2 | NP_067374.2 | |||
| AB043551.1 | 3 | 770 | BAB32728.1 | |
| AB043553.1 | BAB32729.1 | |||
| AB043583.1 | BAB32730.1 | |||
| AF186019.1 | AAF63683.1 | |||
| AK088219.1 | 3 | 715 | ||
| BC019503.1 | AAH19503.1 | |||
| BM950501.1 | 3 | 646 | ||
| BQ442983.1 | 2 | 141 | ||
| BQ745012.1 |
More Information
Set Membership: |
|

Genomic Map