Assay Details
Target Gene Details
Entrez Gene ID: | 207304 |
Gene Name: | HECT domain containing 1 |
Gene Aliases: |
A630086P08Rik, AI844876, b2b327Clo, opm |
Location: |
Chr.12:51743721-51829605 on Build GRCm38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hectd1 | NM_144788.2 | 30 | 5867 | NP_659037.2 |
| AK141970.1 | 8 | 1782 | BAE24902.1 | |
| AK169474.1 | 4 | 1607 | BAE41193.1 | |
| AK173106.1 | 10 | 2396 | BAD32384.1 |
More Information
Set Membership: |
|

Genomic Map