Assay Details
Target Gene Details
Entrez Gene ID: | 207304 |
Gene Name: | HECT domain containing 1 |
Gene Aliases: |
A630086P08Rik, AI844876, b2b327Clo, opm |
Location: |
Chr.12:51743721-51829605 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 38 - Exon 39 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hectd1 | NM_144788.2 | NP_659037.2 | ||
| AK077185.1 | BAC36667.1 | |||
| AK173106.1 | BAD32384.1 |
More Information
Set Membership: |
|

Genomic Map