Assay Details
Target Gene Details
Entrez Gene ID: | 207304 |
Gene Name: | HECT domain containing 1 |
Gene Aliases: |
A630086P08Rik, AI844876, b2b327Clo, opm |
Location: |
Chr.12:51743721-51829605 on Build GRCm38 |
Assay Gene Location: | Within Exon 42 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hectd1 | NM_144788.2 | 42 | 7942 | NP_659037.2 |
| AK077185.1 | 5 | 680 | BAC36667.1 | |
| AK173106.1 | 22 | 4471 | BAD32384.1 | |
| BC010205.1 | 3 | 204 | AAH10205.2 | |
| BC089310.1 | 2 | 113 | AAH89310.1 |
More Information
Set Membership: |
|

Genomic Map