Assay Details
Target Gene Details
Entrez Gene ID: | 211914 |
Gene Name: | ArfGAP with SH3 domain, ankyrin repeat and PH domain 2 |
Gene Aliases: |
6530401G17Rik, Ddef2, Gm1523, Gm592, PAG3, PAP |
Location: |
Chr.12:21111713-21270171 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asap2 | NM_001004364.2 | NP_001004364.2 | ||
| NM_001098168.1 | NP_001091637.1 | |||
| NM_001135192.1 | NP_001128664.1 | |||
| AK147622.1 | BAE28030.1 | |||
| AK147656.1 | BAE28052.1 | |||
| AK172944.1 | BAD32222.1 | |||
| BC096022.1 | AAH96022.1 | |||
| CD351639.1 |

Genomic Map