Assay Details
Target Gene Details
Entrez Gene ID: | 27277 |
Gene Name: | golgi autoantigen, golgin subfamily a, 5 |
Gene Aliases: |
Ret-II |
Location: |
Chr.12:102469134-102497911 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Golga5 | NM_001199004.1 | NP_001185933.1 | ||
| NM_013747.4 | NP_038775.1 | |||
| AB016784.1 | BAA33010.1 | |||
| AF026274.1 | AAF21628.1 | |||
| AK007592.1 | ||||
| AK138455.1 | BAE23668.1 | |||
| AK152533.1 | BAE31289.1 | |||
| AK153010.1 | BAE31649.1 | |||
| AK168765.1 | BAE40601.1 | |||
| BC016883.1 | AAH16883.1 | |||
| BC086782.1 | AAH86782.1 |

Genomic Map