Assay Details
Target Gene Details
Entrez Gene ID: | 58240 |
Gene Name: | HCLS1 binding protein 3 |
Gene Aliases: |
- |
Location: |
Chr.12:8313433-8343824 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hs1bp3 | NM_021429.3 | NP_067404.2 | ||
| AJ132192.1 | CAA10600.1 | |||
| AK040654.1 | ||||
| AK084802.1 | ||||
| AK154696.1 | ||||
| AK170838.1 | BAE42064.1 | |||
| AK212691.1 | ||||
| AK215479.1 | ||||
| AK219357.1 |

Genomic Map