Assay Details
Target Gene Details
Entrez Gene ID: | 104721 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 1 |
Gene Aliases: |
AA409185, DBP-RB |
Location: |
Chr.12:13219307-13249174 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx1 | NM_134040.1 | 17 | 1397 | NP_598801.1 |
| AK028341.1 | 17 | 1394 | BAC25893.1 | |
| AK132157.1 | 17 | 1437 | BAE21002.1 | |
| AK153335.1 | 17 | 1421 | BAE31914.1 | |
| AK160982.1 | 17 | 1398 | BAE36130.1 | |
| AK166146.1 | 17 | 1380 | ||
| AK217868.1 | ||||
| BC008570.1 | 17 | 1375 | AAH08570.1 | |
| BC010624.1 | 17 | 1397 | AAH10624.1 |

Genomic Map