Assay Details
Target Gene Details
Entrez Gene ID: | 238377 |
Gene Name: | G protein-coupled receptor 68 |
Gene Aliases: |
BB131428, Ogr1 |
Location: |
Chr.12:100876682-100908198 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr68 | NM_001177673.1 | NP_001171144.1 | ||
| NM_001177674.1 | NP_001171145.1 | |||
| NM_175493.4 | 2 | 311 | NP_780702.1 | |
| AK036659.1 | BAC29521.1 | |||
| AK087410.1 | BAC39863.1 | |||
| AK154649.1 | BAE32740.1 | |||
| AK155886.1 | BAE33483.1 | |||
| BC118034.1 | ||||
| BC118044.1 | ||||
| BY206757.1 | ||||
| BY247743.1 |
More Information
Set Membership: |
|

Genomic Map