Assay Details
Target Gene Details
Entrez Gene ID: | 104721 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 1 |
Gene Aliases: |
AA409185, DBP-RB |
Location: |
Chr.12:13219307-13249174 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx1 | NM_134040.1 | NP_598801.1 | ||
| AJ223068.1 | ||||
| AK028341.1 | BAC25893.1 | |||
| AK132157.1 | BAE21002.1 | |||
| AK153335.1 | BAE31914.1 | |||
| AK160982.1 | BAE36130.1 | |||
| AK166146.1 | ||||
| BC008570.1 | AAH08570.1 | |||
| BC010624.1 | AAH10624.1 |
More Information
Set Membership: |
|

Genomic Map