Assay Details
Target Gene Details
Entrez Gene ID: | 18263 |
Gene Name: | ornithine decarboxylase, structural 1 |
Gene Aliases: |
ODC |
Location: |
Chr.12:17544873-17551502 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Odc1 | NM_013614.2 | NP_038642.2 | ||
| AK077679.1 | ||||
| AK139610.1 | BAE24082.1 | |||
| AK150283.1 | ||||
| AK150343.1 | ||||
| AK152525.1 | ||||
| AK166093.1 | BAE38566.1 | |||
| AK168219.1 | ||||
| AK169214.1 | ||||
| AK172568.1 | ||||
| BC033264.1 | AAH33264.1 | |||
| BC059826.1 | AAH59826.1 | |||
| BC083122.1 | AAH83122.1 | |||
| EU684749.1 | ||||
| M12330.1 | AAA39846.1 | |||
| M20617.2 | AAA51638.1 | |||
| S64539.1 | AAB27809.1 |
More Information
Set Membership: |
|

Genomic Map