Assay Details
Target Gene Details
Entrez Gene ID: | 66375 |
Gene Name: | dehydrogenase/reductase (SDR family) member 7 |
Gene Aliases: |
2310016E22Rik, 5730564L20Rik, AW061210, Retdsr4, Retsdr4 |
Location: |
Chr.12:72650353-72664828 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dhrs7 | NM_025522.5 | NP_079798.2 | ||
| AK009385.1 | ||||
| AK014100.1 | BAB29156.1 | |||
| BC016189.1 | AAH16189.1 | |||
| CX223953.1 |
More Information
Set Membership: |
|

Genomic Map