Assay Details
Target Gene Details
Entrez Gene ID: | 108156 |
Gene Name: | methylenetetrahydrofolate dehydrogenase (NADP+ dependent), methenyltetrahydrofolate cyclohydrolase, formyltetrahydrofolate synthase |
Gene Aliases: |
Dcs, E430024A07Rik, Mthfd |
Location: |
Chr.12:76255232-76319820 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mthfd1 | NM_138745.2 | 5 | 612 | NP_620084.2 |
| AF364579.1 | 5 | 393 | AAL99692.1 | |
| AK042019.1 | 5 | 492 | BAC31133.2 | |
| AK042958.1 | 5 | 448 | BAC31419.1 | |
| AK088700.1 | 5 | 409 | BAC40513.1 | |
| AK129012.1 | 5 | 546 | ||
| AK133848.1 | 4 | 347 | BAE21884.1 | |
| AK159812.1 | 5 | 424 | BAE35392.1 | |
| AK165951.1 | 5 | 444 | ||
| AK180541.1 | 5 | 330 | ||
| BC008523.1 | 5 | 436 | AAH08523.1 |

Genomic Map