Assay Details
Target Gene Details
Entrez Gene ID: | 192119 |
Gene Name: | dicer 1, ribonuclease type III |
Gene Aliases: |
1110006F08Rik, D12Ertd7e, mKIAA0928 |
Location: |
Chr.12:104687740-104751952 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dicer1 | NM_148948.2 | NP_683750.2 | ||
| AB081470.1 | BAC15765.1 | |||
| AF430845.1 | AAM21495.1 | |||
| AK129241.1 | BAC98051.1 |

Genomic Map