Assay Details
Target Gene Details
Entrez Gene ID: | 217666 |
Gene Name: | L-2-hydroxyglutarate dehydrogenase |
Gene Aliases: |
BC016226 |
Location: |
Chr.12:69690436-69724874 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| L2hgdh | NM_145443.2 | NP_663418.1 | ||
| AK145228.1 | BAE26312.1 | |||
| AK152450.1 | BAE31229.1 | |||
| AK168429.1 | BAE40337.1 | |||
| BC016226.1 | AAH16226.1 |
More Information
Set Membership: |
|

Genomic Map