Assay Details
Target Gene Details
Entrez Gene ID: | 70369 |
Gene Name: | BCL2-associated athanogene 5 |
Gene Aliases: |
1600025G07Rik, 4930405J06Rik |
Location: |
Chr.12:111709488-111713256 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bag5 | NM_027404.2 | NP_081680.1 | ||
| BC037642.1 | AAH37642.1 | |||
| CX243791.1 |

Genomic Map