Assay Details
Target Gene Details
Entrez Gene ID: | 12454 |
Gene Name: | cyclin K |
Gene Aliases: |
AW123198, AW413594, CPR4, CycK |
Location: |
Chr.12:108179446-108203362 on Build GRCm38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccnk | NM_009832.2 | NP_033962.2 | ||
| AK146711.1 | BAE27377.1 | |||
| AK154679.1 | BAE32760.1 | |||
| BC027297.1 | AAH27297.1 |
More Information
Set Membership: |
|

Genomic Map