Assay Details
Target Gene Details
Entrez Gene ID: | 238123 |
Gene Name: | component of oligomeric golgi complex 5 |
Gene Aliases: |
5430405C01Rik, C87247, D18362, GOLTC1, GTC90 |
Location: |
Chr.12:31654868-31937630 on Build GRCm38 |
Assay Gene Location: | Within Intron 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cog5 | NM_001163126.1 | NP_001156598.1 | ||
| AK030656.1 | BAC27065.1 | |||
| BM244396.2 |
Target Gene Details
Entrez Gene ID: | 73389 |
Gene Name: | high mobility group box transcription factor 1 |
Gene Aliases: |
1700058O05Rik, BE629963, C330012F01Rik, C86454 |
Location: |
Chr.12:31926455-31950535 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hbp1 | NM_153198.2 | NP_694878.2 | ||
| NM_177993.3 | NP_818774.2 | |||
| AK006835.1 | ||||
| AK028674.1 | BAC26060.1 | |||
| AK028949.1 | ||||
| AK082770.1 | BAC38611.1 | |||
| AK132503.1 | ||||
| AK133127.1 | BAE21520.1 | |||
| AK139211.1 | BAE23919.1 | |||
| AK142187.1 | BAE24968.1 | |||
| BC026853.1 | AAH26853.1 |
More Information
Set Membership: |
|

Genomic Map