Assay Details
Target Gene Details
Entrez Gene ID: | 70934 |
Gene Name: | RIKEN cDNA 4931403G20 gene |
Gene Aliases: |
- |
Location: |
Chr.12:69843686-69853671 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4931403G20Rik | NR_038172.1 | |||
| NR_038173.1 | ||||
| AK016427.1 |
Target Gene Details
Entrez Gene ID: | 399510 |
Gene Name: | mitogen-activated protein kinase kinase kinase kinase 5 |
Gene Aliases: |
4432415E19Rik, GCKR, KHS, MAPKKKK5 |
Location: |
Chr.12:69803757-69893163 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map4k5 | NM_201519.2 | NP_958927.2 | ||
| AK019468.1 | BAB31739.1 | |||
| AK032123.1 | ||||
| AK053775.1 | BAC35517.1 | |||
| AK084891.1 | BAC39305.1 | |||
| BC040381.1 | AAH40381.2 | |||
| BC048173.1 |
More Information
Set Membership: |
|

Genomic Map