Assay Details
Target Gene Details
Entrez Gene ID: | 12161 |
Gene Name: | bone morphogenetic protein 6 |
Gene Aliases: |
D13Wsu115e, Vgr1 |
Location: |
Chr.13:38345716-38499728 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bmp6 | NM_007556.2 | NP_031582.1 | ||
| AK135770.1 | BAE22651.1 | |||
| BC138593.1 | ||||
| BC138595.1 | ||||
| J04566.1 | AAA40548.1 | |||
| X80992.1 | CAA56917.1 |
More Information
Set Membership: |
|

Genomic Map