Assay Details
Target Gene Details
Entrez Gene ID: | 68021 |
Gene Name: | biphenyl hydrolase-like (serine hydrolase, breast epithelial mucin-associated antigen) |
Gene Aliases: |
2010012D11Rik, 5730533B08Rik, AI115341 |
Location: |
Chr.13:34037593-34074074 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bphl | NM_026512.1 | NP_080788.1 | ||
| AK002907.1 | BAB22447.1 | |||
| AK007606.1 | ||||
| AK008202.1 | BAC25206.1 | |||
| AK017801.1 | ||||
| AK078232.1 | BAC37185.1 | |||
| AK152712.1 | BAE31438.1 | |||
| AK170099.1 | BAE41563.1 | |||
| BC023146.1 | AAH23146.1 | |||
| BC025162.1 | AAH25162.1 |
More Information
Set Membership: |
|

Genomic Map