Assay Details
Target Gene Details
Entrez Gene ID: | 77219 |
Gene Name: | prostaglandin reductase 2 |
Gene Aliases: |
1810016I24Rik, 9130222H03Rik, 9630002F03Rik, AI838763, B830026H24Rik, PGR-2, PRG-2, Zadh1 |
Location: |
Chr.12:84285237-84315832 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptgr2 | NM_001252625.1 | NP_001239554.1 | ||
| NM_001252626.1 | NP_001239555.1 | |||
| NM_029880.3 | NP_084156.2 | |||
| AK020666.1 | ||||
| AK036168.1 | BAC29329.1 | |||
| AK040396.1 | ||||
| AK041027.1 | ||||
| BC021466.1 | AAH21466.1 | |||
| CJ050177.1 | ||||
| CJ137771.1 | 1 | 124 |
More Information
Set Membership: |
|

Genomic Map