Assay Details
Target Gene Details
Entrez Gene ID: | 26448 |
Gene Name: | MOK protein kinase |
Gene Aliases: |
RAGE1, Rage, Stk30 |
Location: |
Chr.12:110807798-110840957 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mok | NM_011973.2 | 11 | 1278 | NP_036103.1 |
| AB022695.1 | 11 | 1278 | BAA81689.1 | |
| BC037614.1 | 6 | 2321 | AAH37614.1 | |
| BC087925.1 | 6 | 717 | AAH87925.1 |
More Information
Set Membership: |
|

Genomic Map