Assay Details
Target Gene Details
Entrez Gene ID: | 56043 |
Gene Name: | aldo-keto reductase family 1, member E1 |
Gene Aliases: |
1810061I10Rik, Akr1e2 |
Location: |
Chr.13:4592490-4609164 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akr1e1 | NM_018859.2 | NP_061347.2 | ||
| AK002507.1 | BAB22152.1 | |||
| AK007924.1 | ||||
| BC012692.1 | AAH12692.1 | |||
| U68535.1 | AAB37274.1 |
More Information
Set Membership: |
|

Genomic Map