Assay Details
Target Gene Details
Entrez Gene ID: | 26950 |
Gene Name: | visinin-like 1 |
Gene Aliases: |
VILIP, Vnsl1 |
Location: |
Chr.12:11325245-11436649 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vsnl1 | NM_012038.4 | NP_036168.1 | ||
| AJ489331.1 | CAD33793.1 | |||
| AK144454.1 | BAE25898.1 | |||
| AK160224.1 | BAE35699.1 | |||
| AY101375.1 | AAM48292.1 | |||
| BC046226.1 | AAH46226.1 | |||
| BY264558.1 | ||||
| D21165.1 | BAA04701.1 | |||
| EU373446.1 |

Genomic Map