Assay Details
Target Gene Details
Entrez Gene ID: | 75516 |
Gene Name: | tetratricopeptide repeat domain 32 |
Gene Aliases: |
1700013G20Rik |
Location: |
Chr.12:9029912-9045118 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttc32 | NM_029321.2 | NP_083597.1 | ||
AK005950.1 | BAB24334.1 | |||
AK078146.1 | BAC37147.1 | |||
AK144458.1 | ||||
BC119273.1 |
